Learn More
Lab-Aids™ Osmosis and Diffusion Kit
Observe diffusion through a membrane in two directions at the same time, observe selective permeability of a membrane and understand how osmosis and diffusion relate to cellular life processes on a cellular level.
Supplier: Lab-Aids™ 22
Includes:
- 30 semi-permeable membranes
- Two bottles each of glucose solution and liquid starch
- 30 containers
- 60 glucose testing strips
- 30 individual worksheets and guides
Description
This kit helps students observe first hand the characteristics of a deferentially permeable membrane. Some substances will pass through the membrane and some won't. Some will actually pass through the membrane in both directions simultaneously. Simple color changes help students visualize this biological-physiological phenomenon. Kit is complete for 30 students.Order Info
Class size: 30 students.
Specifications
Osmosis and Diffusion Kit | |
Teacher's guide with msds, 30 student worksheets and guides, 30 membranes, semi-permeable, 60 glucose testing strips, 30 plastic container, 3 droppers, lugol's solution bottle, glucose solution bottle, liquid starch bottle | |
24 Students |
Study the basic life process function of osmosis and diffusion | |
One 50 minute class period |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.