Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lab-Aids Thin-Layer Chromatography Kit
Develop understanding of thin layer chromatography, use Rf values to establish standards and use Rf standards to identify 'unknowns'.
$261.00
Specifications
Class Size | 24 students |
---|---|
For Use With (Application) | Determine The Purity of a System with Thin Layer Chromatography |
Includes | 12 developing chambers with caps, 72 tlc layers, 4 Bottles of tlc solvent, 2 vials of capillary applicators, drop-controlled bottles of one each: sudan iv, methyl violet, indigo carmine, methyl orange and methyl blue, 2 empty vials, 24 student worksheets |
Includes: Developing chambers with caps (12); TLC layers (72); bottles of TLC solvent (4); vials of capillary applicators (2); drop-controlled bottles of one each: Sudan IV, Methyl Violet, Indigo Carmine, Methyl Orange and Methyl Blue; empty vials (2); student worksheets and guide (24); and an instruction manual with MSDS
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S19347
![]() |
Lab-Aids
17 |
Thin Layer Chromatography Kit |
Each for $261.00
|
|
Description
This inexpensive, simple, and very accurate Lab-Aid illustrates the principles of Thin Layer Chromatography (TLC). The lab techniques involve silica-gel coated plastic slides, indicators, developers and chromatography chambers. Each student runs chromatograms of five different sample dye solutions in order to establish Rf (Rate of flow) standards. They also run chromatograms of two 'unknowns,' which are identified for the teacher in the instruction booklet. This approach gives students an opportunity to appreciate the usefulness of TLC. TLC, which is similar and complementary to paper chromatography, can also be employed to determine the purity of a system. This kit is complete for 24 students and can be performed in a single lab period.Order Info
Class size: 24 students.
Specifications
24 students | |
Determine The Purity of a System with Thin Layer Chromatography | |
12 developing chambers with caps, 72 tlc layers, 4 Bottles of tlc solvent, 2 vials of capillary applicators, drop-controlled bottles of one each: sudan iv, methyl violet, indigo carmine, methyl orange and methyl blue, 2 empty vials, 24 student worksheets |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title