Learn More
Lab Armor™ Single and Double Bead Blocks™ Available on GSA/VA Contract for Federal Government customers only.
Designed for use with any standard commercially available dry bath
Manufacturer:  LAB ARMOR 52200GLD
 View more versions of this product
Catalog No. 10-876-017
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Aluminum
0.5L
3 in.
76mm
64mm
3.25 in.
84mm
-80 to 300°C
Gold
Double bead block
2.5 in.
3.75 in.
95mm
-112 to 572°F