Learn More
MicroBioLogics™ Lab Elite™ CRM Campylobacter lari ATCC™ 35221™†
Used for quality control. Microbiologics™ Lab Elite™ CRM Campylobacter lari ATCC™ 35221™† is self-contained device.
Manufacturer:  MicroBioLogics™ 01132CRM
Catalog No. 12-111-317
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Lyophilized microbe pellet, reservoir of hydrating fluid, inoculating swab (KWIK-STIK format)
This item is not returnable. View return policy
Description
Description
  • Delivered in an easy-to-use KWIK-STIK™ format
  • Traceable to reference culture
  • Highly characterized Campylobacter lari
  • For use in ISO 17025 accredited labs and supported by the highest accreditations in the industry
  • ATCC™ 35221™†
Specifications
Specifications
1
Campylobacter lari
KWIK-STIK™
35221
35°C
FDA Bacteriological Analytical Manual (BAM), ISO/IEC 17025
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
† The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.