Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Free Standing Cylinder Restraints

The Fisherbrand™ Free Standing Cylinder Restraints is a fully welded gas cylinder rack for proper securing of cylinders.

 View more versions of this product

Catalog No. 06-000-501


Add to cart

Includes: Security Chains

Description

Description

  • Frame is 2 in. x 2 in. tubular steel
Specifications

Specifications

45 in.
Cylinders
30 in.
Security Chains
40.6 cm
40.6 cm
114.3 cm
Cylinders
76.2cm
16 in.
16 in.
Cylinder Restraint
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit