Learn More
Reagent World IncSupplier Diversity Partner LAB JACK(ANODIZED ALUM) 4X4 DFS Item
Manufacturer:  Reagent World IncSupplier Diversity Partner 939014
ReagentWorld Lab Jack(anodized aluminum), Plate LxW: 4x4 in. (10x10 cm) Max. Height: 5.5 in. (14.5 cm), Ideal for supporting heating appliance, baths, glassware and other items requiring stable regulation. Nonsparking anodized aluminum plates for maximum safety in the lab.
Catalog No. 50-488-313