Learn More
Reagent World IncSupplier Diversity Partner LAB JACK(ANODIZED ALUM) 8X8 DFS Item
Manufacturer:  Reagent World IncSupplier Diversity Partner 939018
ReagentWorld Lab Jack(anodized aluminum), Plate LxW: 8x8 in. (20x20 cm) Max. Height: 11 in. (28 cm), Ideal for supporting heating appliance, baths, glassware and other items requiring stable regulation. Nonsparking anodized aluminum plates for maximum safety in the lab.
Catalog No. 50-488-315