Learn More
Cole-Parmer Cole-Parmer Fine-Point Permanent Lab Markers, Black; 10/Box
Supplier: Cole-Parmer 0996400
Fine-point fiber tip writes on cold, wet surfaces. Quick-drying ink permanently marks plastic; washes off glass, metal, or porcelain—won’t bleed when subjected to water or alcohol.