Learn More
Thermo Scientific™ Lab Rotators
Rotate flasks, test tube racks, beakers, vials, Petri dishes, microwell plates, culture plates, plastic/glass trays and slides with these lab rotors.
Manufacturer:  Thermo Scientific™ 2309Q
 View more versions of this product
Catalog No. 11-671-50Q
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Rotate flasks, test tube racks, beakers, vials, Petri dishes, microwell plates, culture plates, plastic/glass trays and slides in microbiological, immunological and general clinical applications with Thermo Scientific™ Lab Rotators. Improved drive eccentrics and an upgraded motor ensures stability, enhanced long-term performance and quiet, dependable operation. For use in incubators, warm rooms, environmental chambers and refrigerators.
Features:
  • Two platform sizes available
  • Continuous-duty motor provides smooth, quiet orbit
  • Timer makes it easy to perform time studies; set for continuous or timed
  • Autoclavable white silicone nonslip platform mat cleans easily
  • Variable speed control for gentle or vigorous agitation
Warranty:
  • One year parts, 90 days labor
Description & Specifications
Specifications
40 to 220rpm
4° to 40°C (Operating)
10 lb.
9 in.
23cm
4.4 in.
10.62 in.
31cm
120V
16 lb.
20 to 80%
50w
UL listed
12.38 x 10.62 x 4.4 in. (31 x 27 x 11cm)
Rotators
39.2° to 104°F
1.9cm (0.75 in.)
4.5kg
9 in.
23cm
12.38 in.
11cm
27 in.
1 to 60 min.
7.3kg
50/60Hz
0.4A
120V 50/60Hz
40 to 220rpm
Accessories