Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Actin, Smooth Muscle, Ab-1 Mouse Monoclonal Antibody, 40µg/mL, BSA and azide

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Epredia™ Caspase 8 (FLICE) Ab-3, Mouse Monoclonal Antibody.

$302.00 - $1139.00

Specifications

Antigen Caspase 8 (FLICE) Ab-3
Clone 8CSP03; same as 4.1.20
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
MS1143P0
View Documents
Epredia™
MS1143P0
200μg/mL 100μL
Each for $302.00
Add to cart
 
MS1143P1
View Documents
Epredia™
MS1143P1
200μg/mL 500μL
Each for $724.00
Add to cart
 
MS1143P1ABX
View Documents
Epredia™
MS1143P1ABX
1mg/mL 100μL
Each for $791.00
Add to cart
 
MS1143PABX
View Documents
Epredia™
MS1143PABX
1mg/mL 200μL
Each for $1,139.00
Add to cart
 
MS1143P
View Documents
Epredia™
MS1143P
200μg/mL 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Caspase 8 binds to the death effector domain (DED) of FADD through an analogous DED domain present in tandem in the proform of the caspase 8 protein. Recruitment of caspase 8 to the Fas receptor results in oligomerization of the caspase 8 protein, which in turn drives its autoactivation through “self-cleavage”. Activated caspase 8 then activates other downstream caspases including caspase 9, thereby commiting the cell to undergo apoptosis.

Host Species: Mouse

Clone: 8CSP03; same as 4.1.20

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length human Caspase 8 protein

Molecular Weight: 55 kDa

Positive Control: Jurkat cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Caspase 8 (FLICE) Ab-3
Immunoprecipitation, Western Blot
Unconjugated
Mouse
Apoptosis
Recombinant full length human Caspase 8 protein
8CSP03; same as 4.1.20
Monoclonal
Concentrated
RUO
Primary
IgG1
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit