Thermo Scientific Lab Vision™ Autostainer 360-2D

Stain at least 108 slides per day with this fast, space-saving unit that provides flexible options to meet the workflow needs of any laboratory.

Manufacturer: Thermo Scientific A80500004A

 View more versions of this product

Catalog No. A80500004A

  Check Availability
Add to Cart

Alert

Products on this page may include voltages not considered standard in North America. Please review carefully.


    Specifications

    Height (Metric) 58cm
    Length (Metric) 67cm
    Slide Capacity 1–36 slides/run
    Width (Metric) 76cm
    Weight (Metric) ~54kg
    Voltage 120V
    Description Autostainer 360 120V
    Reagent Capacity Up to 40 reagents/run, 12mL/reagent
    Operating Temperature Ambient

    Stain at least 108 slides per day with Thermo Scientific™ Autostainer 360-2D, a fast, space-saving unit that provides flexible options to meet the workflow needs of any laboratory. Autostainers are renowned for their consistency and reliability and are among the most widely used IHC platforms worldwide.

    Benefits:

    • One to 36 slide capacity, each individually programmable
    • User-specified protocols and choice of reagents
    • Multiple staining runs per day
    • Truly hands-free, walk-away operation with advanced timer functions for overnight use
    • Easy, Windows™ 7 based programming
    • Modular options with up to four units controlled from one PC
    • Complemented by the Thermo Scientific™ PT Module designed for fully automated, simultaneous dewaxing and epitope recovery (The PT Module is available seperately or in a system package).
    • Fast flow software logic enables flexible programming, operator ease of use and control of IHC costs

    System Features:

    • Incorporates 2D Data Matrix Bar Coding for Slides and Reagents
    • Can be configured for research or diagnostic applcations at no additional cost
    • Scans rapidly using high precision label detector technology 
    • Has integrated digital bar code scanner
    • Fully configured 2D data matrix software and printer interface
    • Bar code control of all protocol, reagent and slide operations
    • Preprintable data matrix labels with user-defined autoprograms
    • Adaptable for proprietary 2D-labeled reagents or user-defined reagents/labels
    • Optional individual inactivation of slide and/or reagent bar coding

    Includes:

    • 36-slide Autostainer with 2D matrix bar code reader
    • Computer
    • Monitor
    • Label printer(s)
    • Document printer
    • Three metal reinforced, ABS plastic slide racks
    • Reagent rack Reagent vials and caps
    • Autostainer probe cleaning reagent kit
    • Buffer rinse bottle
    • Wash Buffer, DI water and waste containers
    • Universal Power Supply

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit