Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Bcl-X Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Epredia™ Bcl-X Ab-1, Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS715R1

Catalog No. MS715R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Bcl-X has two isoforms, Bcl-XL (long) a 241 amino acid protein and Bcl-XS (short) a 178 amino acid protein lacking a 63 amino acid domain that is well conserved among members of the Bcl-2 family.

Host Species: Mouse

Clone: 2H12

Isotype: IgG2a

Species Reactivity: Human, Mouse, Rat and Pig. Others not tested.

Epitope: aa 3-14

Immunogen: A synthetic peptide, aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-X protein. This amino acid sequence is shared by human and murine Bcl-X protein.

Molecular Weight: 27kDa

Positive Control: Hodgkin's lymphoma

Cellular Localization: Cytoplasmic and Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not optimum)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Bcl-X Ab-1
Monoclonal
Unconjugated
Mouse
1 mL
Apoptosis
Human, Mouse, Porcine, Rat
IgG2a
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
2H12
Ready-to-use for IHC (UVLP)
A synthetic peptide, aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-X protein. This amino acid sequence is shared by human and murine bcl-X protein.
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit