Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Biotinylated Goat anti-Polyvalent (Ready-To-Use)

Employ a labeled streptavidin-biotin immunoenzymatic antigen detection system with the Thermo Scientific™ Lab Vision™ Biotinylated Goat anti-Polyvalent (Ready-To-Use).

Specifications

Conjugate Biotin
Species Goat
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Stability 18 Months
Storage Requirements 2 - 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
TP060BN
View Documents
Thermo Scientific™
TP060BN
60mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
TP125BN
View Documents
Thermo Scientific™
TP125BN
125mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Employ a labeled streptavidin-biotin immunoenzymatic antigen detection system with the Thermo Scientific™ Lab Vision™ Biotinylated Goat anti-Polyvalent (Ready-To-Use).

This technique involves the sequential incubation of the specimen with an unconjugated primary antibody specific to the target antigen, a biotinylated secondary antibody that reacts with the primary antibody, enzyme-labeled streptavidin, and substrate-chromogen.
  • Species: Goat

  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Conjugate: Biotin

Specifications

Specifications

Biotin
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
2 - 8°C
Goat
18 Months
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit