Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Calponin Ab-1 Mouse Monoclonal Antibody, 200µg/mL, BSA and azide

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Calponin Ab-1, Mouse Monoclonal Antibody.

$197.00 - $695.00

Specifications

Antigen Calponin Ab-1
Clone CALP
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1168P
View Documents Promotion Details
Epredia™
MS1168P
Purified Ab with BSA and Azide 1mL
Each for $695.00
Add to cart
 
MS1168P0
View Documents Promotion Details
Epredia™
MS1168P0
Purified Ab with BSA and Azide 100μL
Each for $197.00
Add to cart
 
MS1168P1ABX
View Documents Promotion Details
Epredia™
MS1168P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $462.00
Add to cart
 
MS1168PABX
View Documents Promotion Details
Epredia™
MS1168PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $586.00
Add to cart
 
MS1168R7
View Documents Promotion Details
Epredia™
MS1168R7
Ready-to-use for IHC (UVLP) 7mL
Each for $403.00
Add to cart
 
MS1168R1
View Documents Promotion Details
Epredia™
MS1168R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
MS1168RQ
View Documents Promotion Details
Epredia™
MS1168RQ
Ready-to-use for IHC (Quanto) 12mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Calponin, a calmodulin, binds tropomyosin and F-actin and is thought to be involved in the regulation of smooth muscle contraction. The expression of calponin is restricted to smooth muscle cells and is a marker of the differentiated contractile phenotype of developing smooth muscle. Two isoforms of calponin exist whose molecular weights are 34 kDa and 29 kDa. Expression of the 29 kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions.

Host Species: Mouse

Clone: CALP

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not tested.

Epitope: Not determined

Immunogen: Crude human uterus extract

Molecular Weight: 34 kDa

Positive Control: Myoepithelial cells in breast ducts

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Calponin Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
CALP
Monoclonal
Mouse
Muscle Biology
Crude human uterus extract
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit