Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Lab Vision™ Calretinin Ab-2 Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Calretinin Ab-2, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS1859R1
Description
Calretinin is an intracellular calcium-binding protein belonging to the troponin C superfamily characterized by a structural motif described as the EF-hand domain. The immunohistochemical detection of calretinin in developing cerebellum is restricted to the later stages indicated by weak staining from week 21 of gestation, in Purkinje and basket cells and in neurons of the dentate nucleus. The intensity of staining increases as the cerebellum matures. In tumors, calretinin has been detected in mesotheliomas and some pulmonary adenocarcinomas.Host Species: Mouse
Clone: DAK Calret 1
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant full-length human calretinin protein cloned from Mero-41 cells
Molecular Weight: 31.5 kDa
Positive Control: Mesothelioma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Note:
USA: RUO; Int'l: RUO
Specifications
Calretinin Ab-2 | |
Monoclonal | |
Unconjugated | |
Recombinant full length human calretinin protein cloned from Mero-41 cells | |
RUO | |
Primary | |
Ready-to-Use |
Immunohistochemistry (Paraffin) | |
DAK Calret 1 | |
Mouse | |
1 mL | |
Oncogenesis | |
Human | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction