Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calretinin Rabbit Polyclonal Antibody, Epredia™

Stain formalin-fixed, paraffin-embedded tissues using Epredia™ Calretinin, Rabbit Polyclonal Antibody.

$679.00

Specifications

Antigen Calretinin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Format Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Format Price Quantity & Availability  
RB9002P1
View Documents
Epredia
RB9002P1
500μL Concentrated
Each for $679.00
Add to cart
 
RB9002P0
View Documents
Epredia
RB9002P0
100μL Concentrated This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB9002P
View Documents
Epredia
RB9002P
1mL Concentrated This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB9002R1
View Documents
Epredia
RB9002R1
1mL Prediluted This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB9002R7
View Documents
Epredia
RB9002R7
7mL Prediluted This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Calretinin is an intracellular calcium-binding protein belonging to the troponin C superfamily characterized by a structural motif described as the EF-hand domain. The immunohistochemical detection of calretinin in developing cerebellum is restricted to the later stages indicated by weak staining from week 21 of gestation, in Purkinje and basket cells and in neurons of the dentate nucleus. The intensity of staining increases as the cerebellum matures. In tumors, calretinin has been detected in mesotheliomas and some pulmonary adenocarcinomas.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length mouse calretinin protein

Molecular Weight: 31.5 kDa

Positive Control: Mesothelioma

Cellular Localization: Nuclear and Cytoplasmic

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Calretinin
Polyclonal
Rabbit
Oncogenesis
Recombinant full-length mouse calretinin protein
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit