Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD105/Endoglin/TGFβ 1/3 Receptor Rabbit Polyclonal Antibody with BSA and azide

Obtain accurate and reliable results in immunohistochemistry experiments with Epredia™ CD105/Endoglin/TGF β 1/3 Receptor, Rabbit Polyclonal Antibody.

Specifications

Antigen CD105/Endoglin/TGF beta 1/3 Receptor
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Format Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Format Price Quantity & Availability  
RB9291P0
View Documents
Epredia™
RB9291P0
100μL Concentrated This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB9291P1
View Documents
Epredia™
RB9291P1
500μL Concentrated This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB9291P
View Documents
Epredia™
RB9291P
1mL Concentrated This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB9291R1
View Documents
Epredia™
RB9291R1
1mL Prediluted This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB9291R7
View Documents
Epredia™
RB9291R7
7mL Prediluted This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

CD105 or Endoglin or GP160 is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists as an O- and N-glycosylated homodimer. Up-regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation-associated endothelial cell marker. CD105 binds to TGF-β1 and β3 with high affinity but not TGF-β2.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested

Epitope: Not known

Immunogen: Recombinant protein encoding aa 26-176 of human CD105

Molecular Weight: 95 kDa (monomer) and 190 kDa (dimer)

Positive Control: Tonsil, Placenta

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD105/Endoglin/TGF beta 1/3 Receptor
Polyclonal
Rabbit
Angiogenesis
Recombinant protein encoding aa 26-176 of human CD105
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit