Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD45RB Ab-3 Mouse Monoclonal Antibody, 200µg/mL, BSA and azide

Target human CD45RB in immunohistochemistry applications with Epredia™ CD45RB Ab-3, Mouse Monoclonal Antibody.

Specifications

Antigen CD45RB Ab-3
Clone PD7/26
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1846R1
View Documents
Epredia™
MS1846R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

CD45RB is expressed on mature B lymphocytes and the majority of lymphomas and leukemias of B-cell origin. This antigen is also expressed on the surface of cells of myeloid lineage and in the cytoplasm of tissue macrophages.

Host Species: Mouse

Clone: PD7/26

Isotype: IgG1/κ

Species Reactivity: Human. Others not known

Epitope: Exon B

Immunogen: Human peripheral blood lymphocytes maintained in T-cell growth factor

Molecular Weight: 170 kDa

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD45RB Ab-3
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG2a, κ
PD7/26
Monoclonal
Murine
Hematopoietic
Human peripheral blood lymphocytes maintained in T cell growth factor
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit