Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD74 Ab-1 Mouse Monoclonal Antibody, 200µg/mL, BSA and azide

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and flow cytometry experiments.

Specifications

Antigen CD74 Ab-1
Clone LN2
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS131R1
View Documents Promotion Details
Epredia™
MS131R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Expression of the invariant chain of the HLA-DR antigen (CD74) by B-lymphocytes begins in the pre-B stage but is lost before the final differentiation to plasma cells.

Host Species: Mouse

Clone: LN2

Isotype: IgG1/κ

Species Reactivity: Human and Baboon. Does not react with rat. Others not known

Epitope: Not determined

Immunogen: SU-DHL-4 lymphoma cells

Molecular Weight: 35 kDa

Positive Control: Raji cells, Tonsil/lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry

Specifications

Specifications

CD74 Ab-1
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IVD
Primary
IgG2a, κ
LN2
Monoclonal
Murine
B-Cell
SU-DHL-4 lymphoma cells
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit