Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Lab Vision™ CD8, Rabbit Polyclonal Antibody
Ensure accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD8, Rabbit Polyclonal Antibody.
Supplier: Epredia™ RB9009R1
Description
CD8 molecule consists of two chains, termed alpha and Beta chain, which are expressed as a disulphide-linked alpha/Beta heterdimer or as an alpha/alpha homodimer on T cell subset, thymocytes and NK cells. The majority of CD8+ T cells express CD8 as alpha/Beta heterdimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 alpha chain (but not Beta chain).Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: C-terminus
Immunogen: Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of α-chain of the CD8 molecule
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD8 | |
Polyclonal | |
Rabbit | |
1 mL | |
T-Cell | |
Human | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of alpha chain of the CD8 molecule | |
IVD | |
Primary | |
Prediluted |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction