Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CEA (Carcinoembryonic Antigen)/CD66e Ab-2 Rabbit Polyclonal Antibody with BSA and azide

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Specifications

Antigen CEA (Carcinoembryonic Antigen)/CD66e Ab-2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 4
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
RB368R1
View Documents Promotion Details
Epredia™
RB368R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB368A0
View Documents Promotion Details
Epredia™
RB368A0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB368A1
View Documents Promotion Details
Epredia™
RB368A1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB368RQ
View Documents Promotion Details
Epredia™
RB368RQ
Ready-to-use for IHC (Quanto) 12mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Carcinoembryonic antigen (CEA), is synthesized during development in the fetal gut, and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. Antibody to CEA is reportedly useful in identifying the origin of various metastatic adenocarcinomas and in distinguishing pulmonary adenocarcinomas (60-70% are CEA+) from pleural mesotheliomas (rarely or weakly CEA+).

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Weakly reacts with Rat. Does not react with dog. Others not known

Immunogen: Human CEA isolated from hepatic metastasis of colon adenocarcinoma

Molecular Weight: 180 kDa

Positive Control: LS174T cells or colon CA

Cellular Localization: Cytoplasmic and luminal membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Specifications

Specifications

CEA (Carcinoembryonic Antigen)/CD66e Ab-2
Polyclonal
Rabbit
Oncogenesis
Human CEA isolated from hepatic metastasis of colon adenocarcinoma
Immunoprecipitation, Western Blot
Unconjugated
IVD
Primary
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit