Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin (Muscle Cell Marker) Rabbit Polyclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Manufacturer:  Epredia RB9014R7

 View more versions of this product

Catalog No. RB9014R7


Add to cart

Description

Description

Desmin is an intermediate filament protein of both smooth and striated muscles. Antibody to desmin reacts with striated (skeletal and cardiac) as well as smooth muscle cells. In skeletal and cardiac muscles, the staining is confined to the Z-bands giving a characteristic striated appearance. Anti-desmin antibody is useful in identification of tumors of myogenic origin.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Near C-terminus

Immunogen: A synthetic peptide mapping near the C-terminus of human desmin

Molecular Weight: 53 kDa

Positive Control: Muscle or Sarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation
Specifications

Specifications

Desmin (Muscle Cell Marker)
Polyclonal
Unconjugated
Rabbit
7 mL
Muscle Biology
Human, Murine
IgG
Immunoprecipitation, Western Blot
Polyclonal
Ready-to-use for IHC (UVLP)
A synthetic peptide mapping near the C-terminus of human desmin
IVD
Primary
Prediluted
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit