Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Fast-Red Substrate System

Employ a widely used chromogen for immunohistochemical staining and immunoblotting Thermo Scientific™ Lab Vision™ Fast-Red Substrate System.

Specifications

Chromagen Type Fast Red Tablets
Substrate Type Naphthol Phosphate Substrate
Storage Requirements Store at 2° to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Substrate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Substrate Quantity Price Quantity & Availability  
TA060AF
View Documents
Thermo Scientific™
TA060AF
60mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
TA125AF
View Documents
Thermo Scientific™
TA125AF
125mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Employ a widely used chromogen for immunohistochemical staining and immunoblotting Thermo Scientific™ Lab Vision™ Fast-Red Substrate System.

In the presence of alkaline phosphatase enzyme, Fast-Red produces a bright red precipitate that is soluble in alcohol.

Includes:

TA-060-AF: 12 Fast-Red Tablets (TA-012-AFC), 60mL Naphthol Phosphate Substrate (TA-060-AFS)

TA-125-AF: 25 Fast-Red Tablets (TA-025-AFC), 125mL Naphthol Phosphate Substrate (TA-125-AFS)

Specifications

Specifications

Fast Red Tablets
Naphthol Phosphate Substrate
Store at 2° to 8°C
Safety and Handling

Safety and Handling

ShelfLife : 18 Months

SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit