Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Flt-4 Rabbit Polyclonal Antibody with BSA and azide

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Epredia™ Flt-4, Rabbit Polyclonal Antibody.

Specifications

Antigen Flt-4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
RB9255R1
View Documents
Epredia™
RB9255R1
Prediluted 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Three cell membrane receptor tyrosine kinases, Flt (also designated VEGF-R1), Flk-1 (also designated VEGF-R2) and Flt-4 (also designated VEGF-R3), putatively involved in the growth of endothelial cells, are characterized by the presence of seven immunoglobulin-like sequences in their extracellular domain. On the basis of structural similarity to Flt and Flk-1, it has been speculated that Flt-4 might represent a third receptor for either VEGF or a VEGF-related ligand.

Host Species: Rabbit

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide derived from C-terminus of human Flt-4

Molecular Weight: 150 kDa

Positive Control: Placenta or Breast CA

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation
Specifications

Specifications

Flt-4
Polyclonal
Rabbit
Angiogenesis
A synthetic peptide derived from C-terminus of human Flt-4
Immunoprecipitation, Western Blot
Unconjugated
IVD
Primary
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit