Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Granulocyte (Myeloid Cell Marker) Ab-1 Mouse Monoclonal Antibody, 200µg/mL, BSA and azide

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

Specifications

Antigen Granulocyte (Myeloid cell Marker) Ab-1
Clone BM-2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS189R1
View Documents
Epredia™
MS189R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Host Species: Mouse

Clone: BM-2

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not known.

Immunogen: Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes

Positive Control: Tonsil or lymph node

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
Specifications

Specifications

Granulocyte (Myeloid cell Marker) Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG1
BM-2
Monoclonal
Murine
Cancer and Tumor Biology
Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit