Learn More
Epredia™ Lab Vision™ Heat Shock Protein 60 (HSP60) Ab-1, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and electron microscopy experiments.
Supplier: Epredia™ MS120R1
Description
A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of heat shock proteins (hsp's) or stress response proteins (srp's). hsp60 is a potential antigen in a number of autoimmune diseases including human arthritis.Host Species: Mouse
Clone: LK1
Isotype: IgG1
Species Reactivity: Human, Mouse, Rat, Hamster and Chicken. Does not react with bacteria, helminths and spinach. Others not known.
Epitope: aa 383-447
Immunogen: Recombinant HSP60 protein
Molecular Weight: 60 kDa
Positive Control: BT474 or MAD109 cells, Breast CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native verified)
- Electron Microscopy
Note:
USA: RUO; Int'l: RUO
Specifications
Heat Shock Protein 60 (HSP60) Ab-1 | |
Monoclonal | |
Unconjugated | |
Mouse | |
1 mL | |
Stress Response | |
Chicken, Hamster, Human, Mouse, Rat | |
IgG1 |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
LK1 | |
Ready-to-use for IHC (UVLP) | |
Recombinant HSP60 protein | |
RUO | |
Primary | |
Prediluted |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.