Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Hepatitis B Virus Core Antigen (HBVcAg) Ab-1 Rabbit Polyclonal Antibody with BSA and azide

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Hepatitis B Virus Core Antigen (HBVcAg) Ab-1, Rabbit Polyclonal Antibody.

Specifications

Antigen Hepatitis B Virus Core Antigen (HBVcAg) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
RB1413R1
View Documents Promotion Details
Epredia™
RB1413R1
Prediluted 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB1413R7
View Documents Promotion Details
Epredia™
RB1413R7
Prediluted 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Hepatitis B virus is spherical in shape with a diameter of 42nm. It contains a 27nm partially double stranded DNA core enclosed within a lipoprotein coat. The antigenic activity of the nucleocapsid core is designated as hepatitis B core antigen. The antigens in the outer surface are called as hepatitis B virus surface antigens. Core antigens are localized within the nuclei whereas the surface antigens are present in the cytoplasm of the infected cells. Antibodies to surface antigens appear in circulation at an early stage of infection whereas the antibodies to the core antigens are detected after several weeks.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Hepatitis B virus

Positive Control: Hepatitis B virus infected liver

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Hepatitis B Virus Core Antigen (HBVcAg) Ab-1
Polyclonal
Rabbit
Infectious Disease
Hepatitis B virus
Immunohistochemistry (Paraffin)
Unconjugated
ASR
Primary
IgG
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit