Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER-2/c-erbB-2/neu Ab-17 Mouse Monoclonal Antibody, Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation (For cytoplasmic), Epredia™ HER-2/c-erbB-2/neu Ab-17, Mouse Monoclonal Antibody provides accurate, reproducible results..

$615.00 - $1073.00

Specifications

Antigen HER-2/c-erbB-2/neu Ab-17
Clone e2-4001 + 3B5
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS730P
View Documents
Epredia
MS730PA
Purified Ab with BSA and Azide 1mL
Each for $937.00
Add to cart
 
MS730P1
View Documents
Epredia
MS730P1A
Purified Ab with BSA and Azide 500μL
Each for $615.00
Add to cart
 
MS730P1ABX
View Documents
Epredia
MS730P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $689.00
Add to cart
 
MS730PABX
View Documents
Epredia
MS730PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $1,073.00
Add to cart
 
MS730P0
View Documents
Epredia
MS730P0A
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
MS730R7
View Documents
Epredia
MS730R7A
Ready-to-use for IHC (UVLP) 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

c-erbB-2 is a receptor tyrosine kinase of the c-erbB family. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein

Host Species: Mouse

Clone: e2-4001 + 3B5

Isotype: IgG1 + IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Cytoplasmic

Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human HER-2 protein (3B5)

Molecular Weight: 185kDa

Positive Control: LS174T cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (For cytoplasmic)

Specifications

Specifications

HER-2/c-erbB-2/neu Ab-17
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ASR, IVD
Primary
IgG1
e2-4001 + 3B5
Monoclonal
Mouse
EGFR Family
Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human c-erbB-2 protein (3B5)
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit