Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappa Light Chain Ab-1 Mouse Monoclonal Antibody, Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Kappa Light Chain Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results. 

$206.00 - $555.00

Specifications

Antigen Kappa Light Chain Ab-1
Clone L1C1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Mouse
View More Specs

Products 10
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS143B
View Documents
Epredia
MS143B
Biotinylated Ab with BSA and Azide 1mL
Each for $555.00
Add to cart
 
MS143B0
View Documents
Epredia
MS143B0
Biotinylated Ab with BSA and Azide 100μL
Each for $206.00
Add to cart
 
MS143B1
View Documents
Epredia
MS143B1
Biotinylated Ab with BSA and Azide 500μL
Each for $404.00
Add to cart
 
MS143P
View Documents
Epredia
MS143P
Purified Ab with BSA and Azide 1mL
Each for $512.00
Add to cart
 
MS143P1
View Documents
Epredia
MS143P1
Purified Ab with BSA and Azide 500μL
Each for $259.00
Add to cart
 
MS143P1ABX
View Documents
Epredia
MS143P1ABX
Ab Without BSA/Azide (Frozen) 100μL
Each for $329.00
Add to cart
 
MS143R7
View Documents
Epredia
MS143R7
Ready-to-use for IHC (UVLP) 7mL
Each for $298.00
Add to cart
 
MS143R1
View Documents
Epredia
MS143R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
MS143P0
View Documents
Epredia
MS143P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
MS143PABX
View Documents
Epredia
MS143PABX
Ab Without BSA/Azide (Frozen) 200μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Antibody to the κ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: L1C1

Isotype: IgG1/k

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: B-lymphoma cells

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Kappa Light Chain Ab-1
Immunohistochemistry (Paraffin)
Mouse
Hematopoietic
B lymphoma cells
L1C1
Monoclonal
IVD
Primary
IgG2a kappa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit