Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LH (Luteinizing Hormone) Ab-1, Mouse Monoclonal Antibody, Epredia™

Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ LH (Luteinizing Hormone) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Specifications

Antigen LH (Luteinizing Hormone) Ab-1
Clone LH01
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Form Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Concentration Quantity Price Quantity & Availability  
MS1448P0
View Documents
Epredia
MS1448P0
Concentrated 200μg/mL 100 μL N/A N/A N/A
MS1448P1
View Documents
Epredia
MS1448P1
Concentrated 200μg/mL 500 μL N/A N/A N/A
MS1448P1ABX
View Documents
Epredia
MS1448P1ABX
Concentrated 1mg/mL 100 μL N/A N/A N/A
MS1448PABX
View Documents
Epredia
MS1448PABX
Concentrated 1mg/mL 200 μL N/A N/A N/A
MS1448R1
View Documents
Epredia
MS1448R1
Prediluted, Ready-to-Use 1 mL N/A N/A N/A
Description

Description

Luteinizing hormone (LH) is a tropic hormone which modulates the secretory activity of other endocrine glands. It is produced in the anterior hypophysis of the pituitary gland. The glycoprotein hormone, LH, like follicle stimulating hormone and thyroid stimulating hormone, is composed of a common α-subunit but also a spesofic β-subunit, which characterizes each of these hormones.

Host Species: Mouse

Clone: LH01

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human LH-β

Molecular Weight: 14kDa (β) and 28kDa (αβ)

Positive Control: Anterior pituitary

Cellular Localization: Cytoplasmic

Ab-1 recognizes the beta subunit both free and intact of luteinizing hormone (LH). Ab-1 is useful for the labeling of normal gonadotropic cells of the pituitary and also for the classification of pituitary adenomas and in the differential identification of primary and metastatic tumors of the pituitary.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

LH (Luteinizing Hormone) Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human
IgG2a κ
LH01
Monoclonal
Mouse
Steroids Hormones and Growth Factors
Purified human LH-Beta
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.