Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Liquid Fast-Red Substrate System

Employ a widely used chromogen for immunohistochemical staining and immunoblotting with the Thermo Scientific™ Lab Vision™ Liquid Fast-Red Substrate System.

Specifications

Chromagen Type Fast Red
Substrate Type Naphthol Phosphate
Storage Requirements Store at 2° to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Substrate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Substrate Quantity Price Quantity & Availability  
TA060AL
View Documents
Thermo Scientific™
TA060AL/DEL
60mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
TA125AL
View Documents
Thermo Scientific™
TA125AL
125mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Employ a widely used chromogen for immunohistochemical staining and immunoblotting with the Thermo Scientific™ Lab Vision™ Liquid Fast-Red Substrate System.

In the presence of alkaline phosphatase enzyme, Fast-Red produces a bright red reaction product.
  • Bright red reaction product can be seen using brightfield or fluorescent microscopy
  • Liquid Fast-Red is not soluble in xylenes and alcohols; therefore it may be permanently mounted

Staining Protocol:

1. Prepare the working solution immediately before use.

2. Add 1 drop (40µL) of Liquid Fast-Red Chromogen to 3mL of Naphthol Phosphate Substrate and mix it well.

3. Apply solution to tissue section. Use working solution within 15 minutes of preparation or decreased sensitivity may result.

4. Incubate tissue section for 10-25 minutes.

5. Rinse tissue, counterstain, and coverslip.

For permanent cover slip mounting:

a. Air dry slides on 37°C warming plate before applying permanent mounting media (preferred method).

OR

b. Dehydrate and clear quickly through alcohol and xylene (no more than 10 seconds each) before mounting with permanent mounting media.

For aqueous cover slip mounting:

a. Any gelatin-based mounting media (i.e., Permafluor TA-030-FM)

b. UltraMount aqueous mounting media (TA-060-UM), dries hard, for use without a cover slip)

c. VisionMount aqueous mounting media (TA-060-UG), dries hard, for use with a cover slip)

Note: remove UltraMount and VisionMount with alcohol.

Specifications

Specifications

Fast Red
Naphthol Phosphate
Store at 2° to 8°C
Safety and Handling

Safety and Handling

ShelfLife : 18 Months

SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit