Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mammaglobin Mouse Monoclonal Antibody, Epredia™
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Mammaglobin, Mouse Monoclonal Antibody.
Specifications
Antigen | Mammaglobin |
---|---|
Clone | 304-1A5 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS1919A1
|
Epredia
MS1919A1 |
Concentrated | 500 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS1919R1
|
Epredia
MS1919R1 |
Prediluted | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS1919RQ
|
Epredia
MS1919RQ |
Ready-to-Use | 12 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Mammaglobin, a member of the uteroglobin family, is a 10kDa glycoprotein. Mammaglobin mRNA expression is breast tissue specific. It is overexpressed in human breast cancer. The use of Mammaglobin RT-PCR has been demonstrated to detect disseminated breast cancer. In a study to evaluate the frequency and specificity of mammaglobin, 80% of all primary and metastatic breast tumors tested were immunopositive irrespective of tumor grade, while mammaglobin mRNA was detected in 60% of the peripheral stem cell collections from breast cancer patients.Host Species: Mouse
Clone: 304-1A5
Isotype: IgG1
Species Reactivity: Human. Others not tested.
Immunogen: Recombinant full-length protein
Molecular Weight: 10kDa
Positive Control: Breast carcinoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Mammaglobin | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
ASR | |
Recombinant full-length protein |
304-1A5 | |
Monoclonal | |
Mouse | |
Cancer and Tumor Biology | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title