Learn More
Epredia™ Lab Vision™ Milk Fat Globule 1 Human (HMFG1) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Epredia™ Milk Fat Globule 1, Human (HMFG1) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Supplier: Epredia™ MS512R1
Description
Human milk fat globule membrane protein (HMFG1) is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as on the outer surface of the human milk fat globule. HMFG1 is considered as a differentiation marker. It is useful as specific breast epithelial marker in normal and neoplastic mammary development.
Host Species: Mouse
Clone: EDM45
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: Delipidated human milk fat globule membrane preparation
Molecular Weight: 40-45kDa
Positive Control: SKBR-3 cells; Breast, colon, or lung tissues
Cellular Localization: Membrane and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Milk Fat Globule 1, Human (HMFG1) Ab-1 | |
Monoclonal | |
Unconjugated | |
Mouse | |
1 mL | |
Cancer and Tumor Biology | |
Human | |
IgG2a κ |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
EDM45 | |
Ready-to-use for IHC (UVLP) | |
Delipidated human milk fat globule membrane preparation | |
RUO | |
Primary | |
Prediluted |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.