Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Oct-2 Rabbit Polyclonal Antibody with BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ Oct-2, Rabbit Polyclonal Antibody.

Supplier:  Epredia™ RB9297R1

 View more versions of this product

Catalog No. RB9297R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Oct-2 is a transcription factor belonging to the POU homeodomain family that binds to the Ig gene octomer sites regulating B cell specific genes. Oct-2 protein expression is not restricted to B cells, although expression levels are much higher in these cells. Oct-2 expression is reported to be significantly greater in germinal center derived lymphomas, although other B cell lymphomas also display high levels of expression.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide derived from C-terminus of human Oct-2

Molecular Weight: ∽80kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

OCT2
Polyclonal
Rabbit
1 mL
Transcription Regulation
Human, Mouse
IgG
Immunohistochemistry (Paraffin)
Unconjugated
Synthetic peptide derived from C-terminus of human Oct-2
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit