Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Pax-5 Rabbit Monoclonal Antibody

Take advantage of part of a comprehensive antibody portfolio with the Epredia™ Pax-5, Rabbit Monoclonal Antibody. 

$856.00 - $2215.00

Specifications

Antigen PAX-5
Clone SP34
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
RM9133RQ
View Documents
Epredia™
RM9133RQ
Ready-to-use for IHC (Quanto) 12mL
Each for $856.00
Add to cart
 
RM9133S
View Documents
Epredia™
RM9133S
Culture Supernatant with Azide 1mL
Each for $2,215.00
Add to cart
 
RM9133R7
View Documents
Epredia™
RM9133R7
Ready-to-use for IHC (UVLP) 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RM9133R1
View Documents
Epredia™
RM9133R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RM9133S0
View Documents
Epredia™
RM9133S0
Culture Supernatant with Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

PAX-5 is a B-cell-specific activator protein (BSAP). In the early stages of B cell developmemt, PAX-5 influences the expression of several B-cell-specific genes, such as CD19 and CD20. PAX-5 is expressed primarily in pro-, pre-, and mature B cells, but not in plasma cells. PAX-5 is expressed in most B-cell malignancies (pre-B and mature B cell lymphomas/leukemias). Over 90% of cases are positive for PAX-5 in Hodgkin's lymphoma, lymphoplasmacytic lymphoma, Merkel cell and small cell carcinoma. T-cell lymphomas, myeloma/plasmacytoma and carcinoid tumors are negative for PAX-5.

Host Species: Rabbit

Clone: SP34

Isotype: IgG

Species Reactivity: Human. Others not tested.

Immunogen: Synthetic peptide derived from the C-terminus of human PAX-5 protein

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

PAX-5
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
SP34
Monoclonal
Rabbit
Cancer and Tumor Biology
Synthetic peptide derived from the C-terminus of human Pax-5 protein
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit