Learn More
PDGFRα Ab-1 Rabbit Polyclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ PDGFRα Ab-1, Rabbit Polyclonal Antibody.
Supplier: Epredia RB1691P0
Description
PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, α and ß coded by two different genes. They possess five immunoglobulin like domains that are involved in ligand binding. Two tyrosine kinase domains are separated by a kinase insert to which PI-3 kinase can bind. PDGF AA can bind autophosphorylate and activate only PDGFR, alpha while PDGF BB can bind and activate to both PDGFR, α and βHost Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, and Rat. Others not known.
Epitope: C-terminal
Immunogen: A synthetic peptide derived from the C-terminus of the precursor form of human PDGFRα
Molecular Weight: ∽190kDa
Postitive Control: Ovarian CA
Cellular Localization: Membrane Specific for PDGFR, α and does not react with PDGFR, β.
Recommended for:
- Immunohistochemistry
Note:
USA: RUO; Int'l: RUO
Specifications
PDGFR, alpha Ab-1 | |
Polyclonal | |
Unconjugated | |
A synthetic peptide derived from the C-terminus of the precursor form of human PDGFR, alpha | |
100 μL | |
Growth Factor | |
Human, Mouse | |
IgG |
Immunohistochemistry (Paraffin) | |
1mg/mL | |
Rabbit | |
Purified with BSA and azide | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.