Learn More
Thermo Scientific™ Lab Vision™ Permanent Fast-Red Quanto
Provide a permanent magenta stain for use with immunohistochemistry procedures.
Specifications
Storage Requirements Store at 2° to 8°C
View More Specs
Products 2
Catalog Number Mfr. No. Substrate Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Substrate Quantity Price Quantity & Availability  
TA060QAL
View Documents
Thermo Scientific™
TA060QAL
60mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
TA125QAL
View Documents
Thermo Scientific™
TA125QAL
125mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Provide a permanent magenta stain for use with immunohistochemistry procedures with the Thermo Scientific™ Lab Vision™ Permanent Fast-Red Quanto. Permanent Fast-Red Quanto produces outstanding results while overcoming typical fast red workflow challanges.
Historically, integrating fast red chromogens into routine Imunohistochemistry was a challenge. Thermo Scientific Permanent Fast-Red Quanto eliminates these challanges and produces the brilliant results you expect.
  • Permanent Results — Does not fade during long term storage
  • Solvent Resistance — Insoluable in xylene or alcohols allowing Fast-Red slides to be coverslipped with routine HEs and IHC slides
  • Automated “On Slide” Chromogen Mixing — The two chromogen reagents are delivered in equal parts and are mixed sequentially, directly on the slide, allowing any open instrument to mix the chromogen on board reducing turn around time and manual steps
  • Fast Incubation Times — With typical incubation times of 11to 15 minutes Permanent Fast Red Quanto is one of the quickest fast reds on the market
Specifications
Specifications
Store at 2° to 8°C
Safety and Handling
Safety and Handling
ShelfLife : 18 Months