Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X)
Employ a wash buffer for immunolabeling techniques with the Epredia™ Lab Vision™ Phosphate Buffered Saline and Tween™ 20 (20X).
$90.00 - $520.00
Specifications
Product Type | PBS and Tween |
---|---|
For Use With (Application) | Anatomical Pathology |
Concentration | 20X |
Content And Storage | Room Temperature |
Description
- For in vitro diagnostic use
- Added to promote more effective washings, resulting in decreased nonspecific background staining
- Aids reagent spreading on slides
Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week.
Specifications
PBS and Tween | |
Anatomical Pathology | |
20X | |
Room Temperature |
For in vitro diagnostic use
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title