Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ PT Module™ Deparaffinization and Heat-Induced Epitope Retrieval Solutions (100X)

Perform Heat Induced Antigen Retrieval (HIER) on formalin fixed paraffin-embedded tissue sections with Thermo Scientific™ Lab Vision™ PT Module™ Epitope Retrieval Solutions.

Specifications

Dilution 100X
Storage Requirements Room Temperature
View More Specs

Products 6
Catalog Number Mfr. No. pH Quantity Price Quantity & Availability  
Catalog Number Mfr. No. pH Quantity Price Quantity & Availability  
TA125PM1X
View Documents
Thermo Scientific™
TA125PM1X
6 125mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
TA125PM2X
View Documents
Thermo Scientific™
TA125PM2X
8 125mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
TA125PM4X
View Documents
Thermo Scientific™
TA125PM4X
9 125mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
TA250PM1X
View Documents
Thermo Scientific™
TA250PM1X
6 250mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
TA250PM2X
View Documents
Thermo Scientific™
TA250PM2X
8 250mL This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
TA250PM4X
View Documents
Thermo Scientific™
TA250PM4X
9 250mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Perform Heat Induced Antigen Retrieval (HIER) on formalin fixed paraffin-embedded tissue sections mounted on glass microscope slides with Thermo Scientific™ Lab Vision™ PT Module™ Deparaffinization and Heat-Induced Epitope Retrieval Solutions.

  • 100X stock solution
  • Dilute 100-fold with distilled water before use
  • Designed for use with Pretreatment Module™
  • Available in Citrate, EDTA or Tris-HCl at various pH levels

Storage Conditions: Store at room temperature. Stable for up to 18 months. Contains no preservatives; for periods longer than three months, store at 2°-8°C.

Specifications

Specifications

100X
Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit