Learn More
Thermo Scientific™ Lab Vision™ Ready-To-Use AEC Substrate System
Utilize 3-Amino-9-Ethylcarbazole (AEC), a widely used chromogen for immunohistochemical staining with the Thermo Scientific™ Lab Vision™ AEC Substrate System. 
Specifications
Chromagen Type 3-Amino-9-Ethylcarbazole (AEC)
Storage Requirements Store at 2° to 8°C
View More Specs
Alert:
The AEC substrate Ready-to-use system contains AEC and Hydrogen Peroxide in N-N-dimethylformamide. AEC is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. Handle with care and dispose of according to regulations.
Products 2
Catalog Number Mfr. No. Chromagen Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Chromagen Quantity Price Quantity & Availability  
TA060SA
View Documents
Thermo Scientific™
TA060SA
60mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
TA125SA
View Documents
Thermo Scientific™
TA125SA
125mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Utilize 3-Amino-9-Ethylcarbazole (AEC), a widely used chromogen for immunohistochemical staining with the Thermo Scientific™ Lab Vision™ AEC Substrate System.
  • Produces an alcohol soluble, red end product and must be used with an aqueous counterstain and mounting media
  • Ready-to-use single solution means all mixing and waste is eliminated
  • Produces a clean, very intense red stain that is free of unwanted background
Specifications
Specifications
3-Amino-9-Ethylcarbazole (AEC)
Store at 2° to 8°C
Safety and Handling
Safety and Handling
ShelfLife : 18 Months