Learn More
Thermo Scientific™ Lab Vision™ Ready-To-Use Streptavidin Peroxidase
Incubate slides for 10 minutes with Thermo Scientific™ Lab Vision™ Streptavidin Peroxidase (Ready-to-Use) reagent for results that will remain stable for 18 months.
Specifications
Description Streptavidin Peroxidase (Ready-To-Use)
Storage Requirements 2° to 8°C. Each component is stable for 18 months.
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
TS060HR
View Documents
Thermo Scientific™
TS060HR
60mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
TS125HR
View Documents
Thermo Scientific™
TS125HR
125mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Incubate slides for 10 minutes with Thermo Scientific™ Lab Vision™ Streptavidin Peroxidase (Ready-to-Use) reagent for results that will remain stable for 18 months. Intended for use in a labeled streptavidin-biotin immunoenzymatic antigen detection system.
Storage Conditions: Store at 2°-8°C.
Specifications
Specifications
Streptavidin Peroxidase (Ready-To-Use)
2° to 8°C. Each component is stable for 18 months.