Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smooth Muscle Actin Rabbit Polyclonal Antibody, Epredia™

Stain smooth muscle cells in vessel walls, gut wall, and myometrium with Epredia™ Actin, Smooth Muscle, Rabbit Polyclonal Antibody.

$658.50

Specifications

Antigen Actin, Smooth Muscle
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Format Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Format Price Quantity & Availability  
RB9010P1
View Documents
Epredia
RB9010P1
500μL Concentrated
Each for $658.50
Add to cart
 
RB9010P0
View Documents
Epredia
RB9010P0
100μL Concentrated This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
RB9010R1
View Documents
Epredia
RB9010R1
1mL Prediluted This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB9010P
View Documents
Epredia
RB9010P
1mL Concentrated This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
RB9010R7
View Documents
Epredia
RB9010R7
7mL Prediluted This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

This antibody stains smooth muscle cells in vessel walls, gut wall, and myometrium. Myoepithelial cells in breast and salivary gland are also stained. It reacts with tumors arising from smooth muscles and myoepithelial cells.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Baboon, Cow, Rabbit, Mouse, Rat and Chicken. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide from N-terminus of human α-smooth muscle isoform of actin

Positive Control: Smooth muscle of arteries, gut wall, uterus and Leiomyoma

Cellular Localization: Cytoplasmic.

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Actin, Smooth Muscle
Polyclonal
Rabbit
Muscle Biology
A synthetic peptide from N-terminus of human alpha-smooth muscle isoform of actin
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit