Learn More
TAG-72/CA 72-4TM Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ TAG-72/CA 72-4 Ab-1, Mouse Monoclonal Antibody.
Manufacturer: Epredia MS138PABX
Description
Tumor-associated glycoprotein (TAG-72) is an oncofetal antigen with properties of a mucin. About 60% of carcinoma patients express TAG-72 in their sera.
Host Species: Mouse
Clone: B72.3
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Dog, Hamster and Rat. Others not known.
Epitope: Mucin-carried-sialylated-Tn epitope
Immunogen: Membrane-enriched fraction of a human breast carcinoma liver metastasis
Molecular Weight: >1000kDa
Positive Control: Breast or lung CA
Cellular Localization: Cell surface and cytoplasmic. The majority of human adenocarcinomas including colorectal, pancreatic, gastric, ovarian, endometrial, mammary, and non-small cell lung cancer display some cell populations that are positive for Ab-1 staining. Weak or no reactivity has been observed with most cell types of normal adult tissue excepting the secretory endometrium. Ab-1 is reportedly useful in distinguishing pulmonary adenocarcinomas (B72.3+) from pleural mesotheliomas (B72.3-).
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
- Radioimaging
Note:
USA: RUO; Int'l: RUO
Specifications
TAG-72/CA 72-4 Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Cancer and Tumor Biology | |
Bovine, Canine, Hamster, Human, Rat | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
B72.3 | |
Unconjugated | |
Membrane-enriched fraction of a human breast carcinoma liver metastasis | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.