Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroid Stimulating Hormone (TSH) Ab-3 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Thyroid Stimulating Hormone (TSH) Ab-3, Mouse Monoclonal Antibody.

Specifications

Antigen Thyroid Stimulating Hormone (TSH) Ab-3
Clone TSH01 + TSH02
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1453R1
View Documents
Epredia
MS1453R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

TSH is a pituitary hormone of 28kDa, which stimulates thyroid growth and production of thyroid hormones.

Host Species: Mouse

Clone: TSH01 + TSH02

Isotype: IgG1/κ + IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human TSH-β

Molecular Weight: 14kDa (β), 28kDa (αβ)

Positive Control: Anterior pituitary

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
Specifications

Specifications

Thyroid Stimulating Hormone (TSH) Ab-3
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG2a, κ
TSH01 + TSH02
Monoclonal
Murine
Steroids Hormones and Growth Factors
Purified human TSH-Beta
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit