Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Trypsin (Enzymatic Pretreatment) Buffer + 30mL Trypsin Concentrate

Epredia™ Lab Vision Trypsin (Enzymatic Pretreatment) is ideal for immunohistochemical procedures.

Specifications

Product Type Trypsin
Storage Requirements Store at 2° to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
TA015TR
View Documents
Epredia™
TA015TR
15mL Buffer + 5mL Trypsin concentrate This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
TA125TR
View Documents
Epredia™
TA125TR
125mL Buffer + 30mL Trypsin concentrate This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Supplied as a two-component system

Storage Conditions: Store at 2°-8°C. Stable for 18 months.

Includes: TA-015-TR: 15mL (Buffer) (TA-015-TRB), 5mL (Trypsin concentrate) (TA-005-TRC)
TA-125-TR: 125mL (Buffer) (TA-125-TRB), 30mL (Trypsin concentrate) (TA-030-TRC)

Specifications

Specifications

Trypsin
Store at 2° to 8°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit