Learn More
Epredia™ Lab Vision™ UltraVision™ Detection System: anti-Polyvalent
A versatile and robust biotin-streptavidin detection system for research applications and cost-conscious laboratories.
Supplier: Epredia™ TP015AF
Description
This technique involves the sequential incubation of the specimen with an unconjugated primary antibody specific to the target antigen, a biotinylated secondary antibody that reacts with the primary antibody, enzyme-labeled streptavidin, and substrate/chromogen.- Labeled streptavidin-biotin immunoenzymatic antigen detection system
Species: Goat
Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Alkaline Phosphatase
Chromogen/Substrate: Fast-Red/Naphthol Phosphate Buffer
Includes:
TP-015-AF: 15mL Ultra V Block (TA-015-UB), 15mL Biotinylated Goat Anti-Polyvalent (TP-015-BN), 15mL Streptavidin Alkaline Phosphatase (TS-015-AP), 15mL Naphthol Phosphate Substrate (TA-015-AFS), three Fast-Red Tablets (TA-003-AFC)
Note:
UltraVision detection system detects a specific antibody bound to an antigen in tissue sections. The specific antibody is located by a biotin-conjugated secondary antibody. This step is followed by the addition of a streptavidin-enzyme conjugate that binds to the biotin present on the secondary antibody. The specific antibody, secondary antibody, and streptavidin-enzyme complex is then visualized with an appropriate substrate/chromogen.
Specifications
Fast-Red | |
Alkaline Phosphatase | |
Goat | |
Naphthol Phosphate Substrate | |
15mL |
75 to 150 Slides | |
Lab Vision™, UltraVision™ | |
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) | |
TP-015-AF: 15mL Ultra V Block (TA-015-UB), 15mL Biotinylated Goat Anti-Polyvalent (TP-015-BN), 15mL Streptavidin Alkaline Phosphatase (TS-015-AP), 15mL Naphthol Phosphate Substrate (TA-015-AFS), three Fast-Red Tablets (TA-003-AFC) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.