Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ UltraVision™ Large Volume Detection System: anti-Polyvalent, HRP (Ready-To-Use)

A versatile and robust biotin-streptavidin detection system for research applications and cost-conscious laboratories.

Manufacturer:  Thermo Scientific™ TP125HL

 View more versions of this product

Catalog No. TP125HL

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Take advantage of a versatile and robust biotin-streptavidin detection system for research applications and cost-conscious laboratories with the Thermo Scientific™ Lab Vision™ UltraVision™ Large Volume Detection System: anti-Polyvalent, HRP (Ready-To-Use).

This technique involves the sequential incubation of the specimen with an unconjugated primary antibody specific to the target antigen, a biotinylated secondary antibody that reacts with the primary antibody, enzyme-labeled streptavidin, and substrate/chromogen.
  • Labeled streptavidin-biotin immunoenzymatic antigen detection system
  • Species: Goat

  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: None provided

Includes:

TP-060-HL: 60mL Ultra V Block (TA-060-UB), 60mL Biotinylated Goat Anti-Polyvalent (TP-060-BN), 60mL Streptavidin Peroxidase (TS-060-HR)

TP-125-HL: 125mL Ultra V Block (TA-125-UB), 125mL Biotinylated Goat Anti-Polyvalent (TP-125-BN), 125mL Streptavidin Peroxidase (TS-125-HR)

Note:

UltraVision detection system detects a specific antibody bound to an antigen in tissue sections. The specific antibody is located by a biotin-conjugated secondary antibody. This step is followed by the addition of a streptavidin-enzyme conjugate that binds to the biotin present on the secondary antibody. The specific antibody, secondary antibody, and streptavidin-enzyme complex is then visualized with an appropriate substrate/chromogen.

Specifications

Specifications

None provided
Peroxidase
125mL
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Store at 2-8°C. Each component is stable for 18 months.
625 to 1250 slides
125mL Ultra V Block (TA-125-UB), 125mL Biotinylated Goat Anti-Polyvalent (TP-125-BN), 125mL Streptavidin Peroxidase (TS-125-HR)
Goat
None provided
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit