Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vascular Endothelial Growth Factor (VEGF) Ab-3 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Vascular Endothelial Growth Factor (VEGF) Ab-3, Mouse Monoclonal Antibody.

Manufacturer:  Epredia MS350P

 View more versions of this product

Catalog No. MS350P

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.

Host Species: Mouse

Clone: JH121

Isotype: IgG1/κ

Species Reactivity: Human and Rabbit. Others not known.

Epitope: Not determined

Immunogen: Recombinant human VEGF121

Molecular Weight: 19-22kDa (reduced)

Positive Control: VEGF165 recombinant protein, Angiosarcoma

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-3 is excellent for neutralizing the bioactivity of human VEGF.

Recommended for:

  • Neutralizes Bioactivity of VEGF

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Vascular Endothelial Growth Factor (VEGF) Ab-3
Monoclonal
200μg/mL
Mouse
1 mL
Angiogenesis
Human, Rabbit
IgG2a κ
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
JH121
Unconjugated
Recombinant human VEGFsubscript121subscript
RUO
Primary
Concentrated
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit