Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Lab Workstation Worksurface

Fisherbrand™ Lab Workstation Worksurface is designed to be used with Fisherbrand™ Lab Workstation.

 View more versions of this product

Catalog No. 06-000-349


Add to cart

Description

Description

  • Choice of 1 in. Thick Black Epoxy or 1 in. Thick Black Phenolic
Specifications

Specifications

600 lb.
Black
76.2cm
2.5cm
Work Surface
182.9cm
272.2kg
30 in.
1 in.
Phenolic
72 in.
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit