Learn More
LabAlliance™ Series I Pump
Easy-maintenance pumps designed for use where pressure above 2500psi and pulse-free operation are not required
Manufacturer:  Scientific Systems, Inc 110SFN01
 View more versions of this product
Catalog No. 06-686-272
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Series I Pump
36.1cm
0.01 to 10mL/min.
16cm
Stainless Steel
110/220V
19.8cm
14.2 in.
110/220V 50/60Hz
6.3 in.
50/60Hz
2500psi
7.8 in.